2-Chloro-p-toluidine

Leverandør: Apollo Scientific
Danger

OR15009-100G OR15009-25G OR15009-500G
APOSOR15009-100GEA 895 NOK
APOSOR15009-100G APOSOR15009-25G APOSOR15009-500G
2-Chloro-p-toluidine
2-Chloro-p-toluidine
Formel: C₇H₈ClN
Kokepunkt: 223…225 °C
Smeltepunkt: 7 °C
Tetthet: 1,151 g/cm³ (25 °C)
Flash punkt: 99 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007666
CAS-nummer: 615-65-6
EINECS: 210-440-7
FN: 3429
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR