5-Bromopyridine-2-carbonyl chloride, Technical Grade

Leverandør: Apollo Scientific
Danger

OR15012-1G
APOSOR15012-1GEA 380 NOK
APOSOR15012-1G
5-Bromopyridine-2-carbonyl chloride, Technical Grade
5-Bromopyridine-2-carbonyl chloride
Formel: C₆H₃BrClNO
Smeltepunkt: 120 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07368290
CAS-nummer: 137178-88-2
FN: 3261
ADR: 8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR