6-Chloro-3-methyl-2-phenyl-4-quinoline carboxylic acid

Leverandør: Apollo Scientific
Warning

OR15394-10G OR15394-1G OR15394-5G
APOSOR15394-10GEA 11000 NOK
APOSOR15394-10G APOSOR15394-1G APOSOR15394-5G
6-Chloro-3-methyl-2-phenyl-4-quinoline carboxylic acid
6-Chloro-3-methyl-2-phenyl-4-quinoline carboxylic acid
Formel: C₁₇H₁₂ClNO₂
MW: 297.74 g/mol
Smeltepunkt: 315 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03419305
CAS-nummer: 154869-06-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR