Naphth-1-ylglyoxal hydrate 95%

Leverandør: Apollo Scientific
Warning

OR1617-100MG
APOSOR1617-100MGEA 340 NOK
APOSOR1617-100MG
Naphth-1-ylglyoxal hydrate 95%
Naphth-1-ylglyoxal hydrate
Formel: C₁₂H₈O₂·H₂O
Kokepunkt: 401 °C (760 mm Hg)
Flash punkt: 196 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08705867
CAS-nummer: 16208-20-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR