Diphenyl phosphate

Leverandør: Apollo Scientific
Warning

Synonymer: Diphenyl hydrogen phosphate

OR16436-100G OR16436-25G OR16436-5G
APOSOR16436-100GEA 1025 NOK
APOSOR16436-100G APOSOR16436-25G APOSOR16436-5G
Diphenyl phosphate
Diphenyl phosphate
Formel: C₁₂H₁₁O₄P
MW: 250.19 g/mol
Smeltepunkt: 62…70 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003033
CAS-nummer: 838-85-7
EINECS: 212-357-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR