2-(Piperazin-1-yl)benzaldehyde

Leverandør: Apollo Scientific
Warning

OR1644-5G
APOSOR1644-5GEA 3675 NOK
APOSOR1644-5G
2-(Piperazin-1-yl)benzaldehyde
2-(Piperazin-1-yl)benzaldehyde
Formel: C₁₁H₁₄N₂O
Lagringstemperatur: Romtemperatur
CAS-nummer: 736991-52-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR