Diphenyl-N-cyanocarbonimidate 97%

Leverandør: Apollo Scientific

Synonymer: Diphenyl N-cyanocarbonimidate, N-Cyanocarbonimidic Acid Diphenyl Ester

OR16450-25G
APOSOR16450-25GEA 1375 NOK
APOSOR16450-25G
Diphenyl-N-cyanocarbonimidate 97%
Diphenyl-N-cyanocarbonimidate
Formel: C₁₄H₁₀N₂O₂
MW: 238.25 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010380
CAS-nummer: 79463-77-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR