(R)-(-)-Phencyphos hydrate

Leverandør: Apollo Scientific
Warning

OR16849-1G OR16849-500MG
APOSOR16849-1GEA 2225 NOK
APOSOR16849-1G APOSOR16849-500MG
(R)-(-)-Phencyphos hydrate
(R)-(-)-Phencyphos hydrate
Formel: C₁₁H₁₇O₅P
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150741
CAS-nummer: 953776-24-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR