4,6-Dioxo-6-phenylhexanoic acid

Leverandør: Apollo Scientific

OR16866-500MG
APOSOR16866-500MGEA 4150 NOK
APOSOR16866-500MG
4,6-Dioxo-6-phenylhexanoic acid
4,6-Dioxo-6-phenylhexanoic acid
Formel: C₁₂H₁₂O₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD12025948
CAS-nummer: 114150-57-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR