4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) 98%

Leverandør: Apollo Scientific

OR17545-5G OR17545-25G
APOSOR17545-5GEA 520 NOK
APOSOR17545-5G APOSOR17545-25G
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) 98%
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₁₂H₂₁NO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03453262
CAS-nummer: 130309-46-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR