4-Chloro-o-toluidine

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Amino-5-chlorotoluene, 4-Chloro-2-methylaniline, 4-Chloro-o-toluidine, Fast Red TR Base

OR17861-100G OR17861-250G OR17861-25G
APOSOR17861-100GEA 645 NOK
APOSOR17861-100G APOSOR17861-250G APOSOR17861-25G
4-Chloro-o-toluidine
4-Chloro-o-toluidine
Formel: C₇H₈ClN
Kokepunkt: 241 °C
Smeltepunkt: 24…30 °C
Tetthet: 1,19 g/cm³ (25 °C)
Flash punkt: 124 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007842
CAS-nummer: 95-69-2
EINECS: 202-441-6
FN: 2239
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR