2,4-Dimethyl-6-hydroxypyrimidine

Leverandør: Apollo Scientific
Warning

Synonymer: 2,6-Dimethylpyrimidin-4-ol, 2,6-Dimethyl-4-pyrimidinol, 6-Hydroxy-2,4-dimethylpyrimidine

OR18376-25G
APOSOR18376-25GEA 750 NOK
APOSOR18376-25G
2,4-Dimethyl-6-hydroxypyrimidine
2,4-Dimethyl-6-hydroxypyrimidine
Formel: C₆H₈N₂O
MW: 124.14 g/mol
Smeltepunkt: 198…200 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006105
CAS-nummer: 6622-92-0
EINECS: 229-577-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR