5-Chloro-6-methoxypyridine-3-carboxylic acid 96%

Leverandør: Apollo Scientific
Warning

Synonymer: 5-Chloro-6-methoxynicotinic acid

OR200033-1G
APOSOR200033-1GEA 4275 NOK
APOSOR200033-1G
5-Chloro-6-methoxypyridine-3-carboxylic acid 96%
5-Chloro-6-methoxypyridine-3-carboxylic acid
Formel: C₇H₆ClNO₃
MW: 187.58 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04972405
CAS-nummer: 884494-85-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR