Methyl-4-benzylmorpholine-2-carboxylate

Leverandør: Apollo Scientific
Warning

Synonymer: Methyl 4-benzylmorpholine-2-carboxylate

OR200057-1G
APOSOR200057-1GEA 0 NOK
APOSOR200057-1G
Methyl-4-benzylmorpholine-2-carboxylate
Methyl-4-benzylmorpholine-2-carboxylate
Formel: C₁₃H₁₇NO₃
MW: 235,29 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD16990656
CAS-nummer: 135782-29-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR