Nicotinic acid hydrazide

Leverandør: Apollo Scientific
Warning

Synonymer: Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide

OR21092-100G OR21092-25G
APOSOR21092-100GEA 750 NOK
APOSOR21092-100G APOSOR21092-25G
Nicotinic acid hydrazide
Nicotinic acid hydrazide
Formel: C₆H₇N₃O
MW: 137.14 g/mol
Smeltepunkt: 159…163 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006383
CAS-nummer: 553-53-7
EINECS: 209-041-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR