3-Chloropyridazine hydrochloride

Leverandør: Apollo Scientific
Warning

OR2124-1G OR2124-5G
APOSOR2124-1GEA 1275 NOK
APOSOR2124-1G APOSOR2124-5G
3-Chloropyridazine hydrochloride
3-Chloropyridazine hydrochloride
Formel: C₄H₃ClN₂·ClH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD12547058
CAS-nummer: 856847-77-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR