4-Bromo-o-toluidine

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Amino-5-bromotoluene, 4-Bromo-2-methylaniline

OR21308-100G OR21308-25G OR21308-500G
APOSOR21308-100GEA 520 NOK
APOSOR21308-100G APOSOR21308-25G APOSOR21308-500G
4-Bromo-o-toluidine
4-Bromo-o-toluidine
Formel: C₇H₈BrN
Kokepunkt: 240 °C
Smeltepunkt: 56…60 °C
Flash punkt: >110 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007825
CAS-nummer: 583-75-5
FN: 3335
ADR: 9,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR