6-Chloro-4-nitro-o-cresol

Leverandør: Apollo Scientific
Warning

Synonymer: 2-Chloro-6-methyl-4-nitrophenol

OR21328-1G
APOSOR21328-1GEA 4000 NOK
APOSOR21328-1G
6-Chloro-4-nitro-o-cresol
6-Chloro-4-nitro-o-cresol
Formel: C₇H₆ClNO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00047740
CAS-nummer: 4102-84-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR