3-Aminothieno[2,3-c]pyridine

Leverandør: Apollo Scientific
Warning

Synonymer: Thieno[2,3-c]pyridin-3-amine

OR2143-250MG
APOSOR2143-250MGEA 3150 NOK
APOSOR2143-250MG
3-Aminothieno[2,3-c]pyridine
3-Aminothieno[2,3-c]pyridine
Formel: C₇H₆N₂S
MW: 150,201 g/mol
Smeltepunkt: 148…150 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09998716
CAS-nummer: 63326-75-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR