3-Methyl-N-[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridinium chloride

Leverandør: Apollo Scientific
Warning

Synonymer: N-[(1E)-3-Oxo-3-phenylprop-1-en-1-yl]-3-picolinium chloride

OR22247-500MG
APOSOR22247-500MGEA 360 NOK
APOSOR22247-500MG
3-Methyl-N-[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridinium chloride
3-Methyl-N-[(1E)-3-oxo-3-phenylprop-1-en-1-yl]pyridinium chloride
Formel: C₁₅H₁₄NO·Cl
MW: 259,731 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00204657
CAS-nummer: 19518-26-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR