3,4,9,10-Perylenetetracarboxylic dianhydride 95%

Leverandør: Apollo Scientific

Synonymer: Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA

OR22484-500G
APOSOR22484-500GEA 2125 NOK
APOSOR22484-500G
3,4,9,10-Perylenetetracarboxylic dianhydride 95%
3,4,9,10-Perylenetetracarboxylic dianhydride
Formel: C₂₄H₈O₆
MW: 392.32 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006916
CAS-nummer: 128-69-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR