syn-Pyridine-2-carboxaldoxime

Leverandør: Apollo Scientific
Warning

Synonymer: syn-2-Pyridinealdoxime

OR24107-100G OR24107-25G OR24107-5G
APOSOR24107-100GEA 1550 NOK
APOSOR24107-100G APOSOR24107-25G APOSOR24107-5G
syn-Pyridine-2-carboxaldoxime
syn-Pyridine-2-carboxaldoxime
Formel: C₆H₆N₂O
Smeltepunkt: 110…112 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167166
CAS-nummer: 1193-96-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR