3-(4-Nitrophenyl)adamantane-1-carboxylic acid 97%

Leverandør: Apollo Scientific
Warning

OR2439-250MG
APOSOR2439-250MGEA 340 NOK
APOSOR2439-250MG
3-(4-Nitrophenyl)adamantane-1-carboxylic acid 97%
3-(4-Nitrophenyl)adamantane-1-carboxylic acid
Formel: C₁₇H₁₉NO₄
MW: 301.35 g/mol
Smeltepunkt: 197…200 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00188065
CAS-nummer: 7123-76-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR