4-Bromothiophene-2-acetic acid

Leverandør: Apollo Scientific
Warning

Synonymer: (4-Bromothien-2-yl)acetic acid, 2-(4-Bromothiophen-2-yl)acetic acid, 4-Bromo-2-(carboxymethyl)thiophene

OR2457-250MG
APOSOR2457-250MGEA 270 NOK
APOSOR2457-250MG
4-Bromothiophene-2-acetic acid
4-Bromothiophene-2-acetic acid
Formel: C₆H₅BrO₂S
MW: 221,08 g/mol
Smeltepunkt: 87…92 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD09839310
CAS-nummer: 161942-89-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR