Diphenyl sulphide 98%

Leverandør: Apollo Scientific
Warning

Synonymer: Diphenylsulphane

OR2458-100G OR2458-25G OR2458-500G
APOSOR2458-100GEA 305 NOK
APOSOR2458-100G APOSOR2458-25G APOSOR2458-500G
Diphenyl sulphide 98%
Diphenyl sulphide
Formel: C₁₂H₁₀S
Kokepunkt: 296 °C
Smeltepunkt: –40 °C
Tetthet: 1,118 g/cm³ (25 °C)
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003064
CAS-nummer: 139-66-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR