5-Amino-4-cyano-1-phenyl-1H-pyrazole 95%

Leverandør: Apollo Scientific
Warning

Synonymer: 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

OR2625T-250MG
APOSOR2625T-250MGEA 270 NOK
APOSOR2625T-250MG
5-Amino-4-cyano-1-phenyl-1H-pyrazole 95%
5-Amino-4-cyano-1-phenyl-1H-pyrazole

Chemiluminescent with antifilarial activity & affinity for the adenosine receptor.

Formel: C₁₀H₈N₄
MW: 184,20 g/mol
Smeltepunkt: 137…139 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00020730
CAS-nummer: 5334-43-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR