cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid

Leverandør: Apollo Scientific
Warning

Synonymer: (1R,2S)-2-(Boc-amino)cyclopentanecarboxylic Acid, cis-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

OR2627-1G
APOSOR2627-1GEA 1750 NOK
APOSOR2627-1G
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
Formel: C₁₁H₁₉NO₄
Smeltepunkt: 90…94 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00800539
CAS-nummer: 136315-70-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR