6-Chloro-m-toluidine 97%

Leverandør: Apollo Scientific
Warning

OR2643-1G
APOSOR2643-1GEA 270 NOK
APOSOR2643-1G
6-Chloro-m-toluidine 97%
6-Chloro-m-toluidine
Formel: C₇H₈ClN
MW: 141.6 g/mol
Kokepunkt: 228…230 °C
Smeltepunkt: 29…30 °C
Flash punkt: 107 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007674
CAS-nummer: 95-81-8
EINECS: 202-454-7
FN: 2239
ADR: 6.1 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR