4'-Hydroxy-3'-methoxyacetophenone

Leverandør: Apollo Scientific
Warning

Synonymer: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin

OR26691-25G
APOSOR26691-25GEA 305 NOK
APOSOR26691-25G
4'-Hydroxy-3'-methoxyacetophenone
4'-Hydroxy-3'-methoxyacetophenone
Formel: C₉H₁₀O₃
MW: 166.18 g/mol
Kokepunkt: 263…265 °C
Smeltepunkt: 112…115 °C
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008747
CAS-nummer: 498-02-2
Merck Indeks: 13,00746

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR