3-Nitro-o-cresol 98%

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Methyl-3-nitrophenol, 1-Hydroxy-2-methyl-3-nitrobenzene

OR27719-25G
APOSOR27719-25GEA 630 NOK
APOSOR27719-25G
3-Nitro-o-cresol 98%
3-Nitro-o-cresol
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Smeltepunkt: 146…152 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007241
CAS-nummer: 5460-31-1
EINECS: 226-739-0
FN: 2446
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR