3-(Chloromethyl)benzoyl chloride 93%

Leverandør: Apollo Scientific
Danger

OR2797-25G
APOSOR2797-25GEA 2100 NOK
APOSOR2797-25G
3-(Chloromethyl)benzoyl chloride 93%
3-(Chloromethyl)benzoyl chloride
Formel: C₈H₆Cl₂O
MW: 189.04 g/mol
Kokepunkt: 149…150 °C/20mmHg
Smeltepunkt: 4 °C
Tetthet: 1.33 g/cm³ (25 °C)
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000682
CAS-nummer: 63024-77-1
EINECS: 263-799-7
FN: 3265
ADR: 8 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR