4-Amino-N-methylbenzamide 98%

Leverandør: Apollo Scientific
Warning

OR2810-10KG
APOSOR2810-10KGEA 0 NOK
APOSOR2810-10KG
4-Amino-N-methylbenzamide 98%
4-Amino-N-methylbenzamide
Formel: C₈H₁₀N₂O
MW: 150,1778 g/mol
Smeltepunkt: 181 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00665792
CAS-nummer: 6274-22-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR