1H-1,2,4-Triazole-3-thiol

Leverandør: Apollo Scientific
Warning

OR28670-100G OR28670-25G OR28670-500G
APOSOR28670-100GEA 360 NOK
APOSOR28670-100G APOSOR28670-25G APOSOR28670-500G
1H-1,2,4-Triazole-3-thiol
1H-1,2,4-Triazole-3-thiol
Formel: C₂H₃N₃S
Smeltepunkt: 217…224 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005229
CAS-nummer: 3179-31-5
EINECS: 221-656-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR