3-tert-Butylbenzoic acid

Leverandør: Apollo Scientific
Warning

Synonymer: 3-(tert-Butyl)benzoic acid

OR2870-5G
APOSOR2870-5GEA 3050 NOK
APOSOR2870-5G
3-tert-Butylbenzoic acid
3-tert-Butylbenzoic acid
Formel: C₁₁H₁₄O₂
MW: 178.23 g/mol
Smeltepunkt: 129…130 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01463861
CAS-nummer: 7498-54-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR