3-(3-Methyl-1H-indol-1-yl)propanoic acid

Leverandør: Apollo Scientific
Warning

OR29133-1G
APOSOR29133-1GEA 2900 NOK
APOSOR29133-1G
3-(3-Methyl-1H-indol-1-yl)propanoic acid
3-(3-Methyl-1H-indol-1-yl)propanoic acid
Formel: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Smeltepunkt: 84…85 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00052088
CAS-nummer: 57662-47-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR