2,2':5',2''-Terthiophene-5-carboxaldehyde

Leverandør: Apollo Scientific

OR29237-5G
APOSOR29237-5GEA 2200 NOK
APOSOR29237-5G
2,2':5',2''-Terthiophene-5-carboxaldehyde
2,2':5',2''-Terthiophene-5-carboxaldehyde
Formel: C₁₃H₈OS₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00115181
CAS-nummer: 7342-41-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR