Ethyl [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate

Leverandør: Apollo Scientific

Synonymer: Ethyl 1,2,4-Triazolo[4,3-a]pyrazine-3-carboxylate

OR300025-1G
APOSOR300025-1GEA 985 NOK
APOSOR300025-1G
Ethyl [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate
Ethyl [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate
Formel: C₈H₈N₄O₂
MW: 192,18 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09909674
CAS-nummer: 723286-67-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR