2-(Bromomethyl)-1-methoxy-3-nitrobenzene

Leverandør: Apollo Scientific
Danger

OR300181-250MG
APOSOR300181-250MGEA 1050 NOK
APOSOR300181-250MG
2-(Bromomethyl)-1-methoxy-3-nitrobenzene
2-(Bromomethyl)-1-methoxy-3-nitrobenzene
Formel: C₈H₈BrNO₃
MW: 246,058 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD17168655
CAS-nummer: 19689-86-2
FN: 3261
ADR: 8,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR