3-Methoxy-o-toluidine

Leverandør: Apollo Scientific

Synonymer: 3-Methoxy-2-methylaniline

OR301008-100G
APOSOR301008-100GEA 5400 NOK
APOSOR301008-100G
3-Methoxy-o-toluidine
3-Methoxy-o-toluidine
Formel: C₈H₁₁NO
MW: 137.1800 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06412568
CAS-nummer: 19500-02-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR