Ethyl-3-cyclopropyl-3-oxopropanoate 95%

Leverandør: Apollo Scientific
Warning

Synonymer: Ethyl 3-cyclopropyl-3-oxopropanoate

OR301083-25G
APOSOR301083-25GEA 270 NOK
APOSOR301083-25G
Ethyl-3-cyclopropyl-3-oxopropanoate 95%
Ethyl-3-cyclopropyl-3-oxopropanoate
Formel: C₈H₁₂O₃
MW: 156,18 g/mol
Kokepunkt: 80…82 °C/6mmHg
Tetthet: 1,058 g/cm³ (25 °C)
Flash punkt: >96 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00184570
CAS-nummer: 24922-02-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR