3-Methoxyazetidine hydrochloride

Leverandør: Apollo Scientific
Warning

OR301086-1G OR301086-5G
APOSOR301086-1GEA 270 NOK
APOSOR301086-1G APOSOR301086-5G
3-Methoxyazetidine hydrochloride
3-Methoxyazetidine hydrochloride
Formel: C4H10ClNO
Smeltepunkt: 143…149 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06804514
CAS-nummer: 148644-09-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR