2,5-Dibromo-3,4-pyridinediamine

Leverandør: Apollo Scientific
Warning

Synonymer: 2,5-Dibromopyridine-3,4-diamine

OR302039-1G
APOSOR302039-1GEA 270 NOK
APOSOR302039-1G
2,5-Dibromo-3,4-pyridinediamine
2,5-Dibromo-3,4-pyridinediamine
Formel: C₅H₅Br₂N₃
MW: 266,921 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD15526814
CAS-nummer: 221241-11-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR