4-Bromo-N-methylpyridin-2-amine

Leverandør: Apollo Scientific

OR302165-1G
APOSOR302165-1GEA 1050 NOK
APOSOR302165-1G
4-Bromo-N-methylpyridin-2-amine
4-Bromo-N-methylpyridin-2-amine
Formel: C₆H₇BrN₂
MW: 187,037 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09834845
CAS-nummer: 468718-65-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR