Azetidine-3-carbonitrile

Leverandør: Apollo Scientific
Warning

OR302435-1G
APOSOR302435-1GEA 7075 NOK
APOSOR302435-1G
Azetidine-3-carbonitrile
Azetidine-3-carbonitrile
Formel: C₄H₆N₂
MW: 82,1 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 732976-86-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR