4-Ethylphenylboronic acid pinacol ester

Leverandør: Apollo Scientific

Synonymer: 2-(4-Ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR303559-5G
APOSOR303559-5GEA 1075 NOK
APOSOR303559-5G
4-Ethylphenylboronic acid pinacol ester
4-Ethylphenylboronic acid pinacol ester
Formel: C₁₄H₂₁BO₂
MW: 232.126 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18643353
CAS-nummer: 1075719-87-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR