2,2-Dimethylcyclobutan-1-ol

Leverandør: Apollo Scientific

OR304366-1G
APOSOR304366-1GEA 23700 NOK
APOSOR304366-1G
2,2-Dimethylcyclobutan-1-ol
2,2-Dimethylcyclobutan-1-ol
Formel: C₆H₁₂O
MW: 100.159 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD26959233
CAS-nummer: 35301-44-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR