3-(Benzyloxy)-2,2-dimethylcyclobutan-1-ol

Leverandør: Apollo Scientific

OR304374-1G
APOSOR304374-1GEA 14125 NOK
APOSOR304374-1G
3-(Benzyloxy)-2,2-dimethylcyclobutan-1-ol
3-(Benzyloxy)-2,2-dimethylcyclobutan-1-ol
Formel: C₁₃H₁₈O₂
MW: 206.281 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD21756139
CAS-nummer: 17139-85-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR