Spiro[4.4]nonane-4,9-dione

Leverandør: Apollo Scientific

OR304402-1G
APOSOR304402-1GEA 26550 NOK
APOSOR304402-1G
Spiro[4.4]nonane-4,9-dione
Spiro[4.4]nonane-4,9-dione
Formel: C₉H₁₂O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00457280
CAS-nummer: 27723-43-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR