2-Bromo-5-phenyl-1,3,4-oxadiazole

Leverandør: Apollo Scientific

OR305588-1G
APOSOR305588-1GEA 1150 NOK
APOSOR305588-1G
2-Bromo-5-phenyl-1,3,4-oxadiazole
2-Bromo-5-phenyl-1,3,4-oxadiazole
Formel: C₈H₅BrN₂O
MW: 225,04 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06660932
CAS-nummer: 51039-53-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR