4-Chlorothieno[3.2.c]pyridine 95%

Leverandør: Apollo Scientific
Warning

Synonymer: 4-Chlorothieno[3,2-c]pyridine

OR30584-1G
APOSOR30584-1GEA 1250 NOK
APOSOR30584-1G
4-Chlorothieno[3.2.c]pyridine 95%
4-Chlorothieno[3.2.c]pyridine
Formel: C₇H₄ClNS
MW: 169,63 g/mol
Smeltepunkt: 97…100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01861830
CAS-nummer: 27685-94-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR